missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydrochloric Acid, 25% (v/v), Ricca Chemical
Supplier: Ricca Chemical Company 357716
Specifications
Hydrochloric Acid | |
7647-01-0,7732-18-5 | |
25% (v/v) | |
Natural Poly Bottle | |
500 mL | |
Liquid | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 |
Colorless to Green-Yellow | |
76.0,22.32 | |
79.0,23.24 | |
Aqueous Solution | |
ClH | |
MFCD00011324 MFCD00792839 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 |
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
hydrogen chloride |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 | |
Cl |