missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric Acid, 25% (v/v) Aqueous Solution, Ricca Chemical

$62.48 - $84.52

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.458
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name chlorane
SMILES Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
3577-16
View Documents
Ricca Chemical Company
RI357716
500mL Poly natural
Each for $62.48
 
3577-32
SDS View Documents
Ricca Chemical Company
RI357732
1L Poly natural
Each for $84.52
 
Description

Description

Specifications

Chemical Identifiers

7647-01-0
36.458
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt
CHEBI:17883
Cl
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
chlorane

Specifications

25% (v/v)
500mL
76.0,22.32
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
313
Liquid
Aqueous Solution
Colorless to Green-Yellow
7647-01-0,7732-18-5
79.0,23.24
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt
Cl
36.458
CHEBI:17883
Poly natural
Hydrochloric Acid
SDS