Please login to your online account to display your discounted pricing

Hydrochloric Acid, 25% v/v (1+3), Fisher Chemical


Standardized at 25°C to pH 8.2 against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate

Quantity 4L
Packaging Poly Bottle

Manufacturer: fisher scientific company  SA8124

 View more versions of this product

Catalog No. SA812-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid, 25% (v/v) (1+3)
Color Clear
Quantity 4L
pH 1
Cleanup Symbol Acid
CAS 7732-18-5
ChemAlert Storage Symbol White
CAS 7647-01-0
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Molecular Formula HCl·H2O
MDL Number MFCD00011324
Concentration 2.96 to 3.03N
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure 5.7mmHg at 0°C
NIST Traceable to NIST.
Melting Point −74°C
Boiling Point 81.5°C

Specifications

Chemical Name or Material Hydrochloric Acid, 25% (v/v) (1+3)
CAS 7732-18-5
CAS 7647-01-0
Molecular Formula HCl·H2O
MDL Number MFCD00011324
Accessories
   

SDS & Safety Documents