Please login to your online account to display your discounted pricing

Hydrochloric Acid, 25% v/v (1+3), Fisher Chemical

Standardized at 25°C to pH 8.2 against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate

$130.08 - $1,601.38

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid, 25% (v/v) (1+3)
CAS 7732-18-5
CAS 7647-01-0
Molecular Formula HCl·H2O
MDL Number MFCD00011324
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SA812-20

  SDS
fisher scientific company
SA81220
PolyPac™ 20L Each for $1,601.38

SA812-1

  SDS
fisher scientific company
SA8121
Poly Bottle 1L Each for $130.08
Case of 6 Each for $780.48

SA812-4

  SDS
fisher scientific company
SA8124
Poly Bottle 4L Each for $320.23
Case of 4 Each for $1,280.92
Description & Specifications

Specifications

Chemical Name or Material Hydrochloric Acid, 25% (v/v) (1+3)
Color Clear
Quantity 20L
pH 1
Cleanup Symbol Acid
CAS 7732-18-5
ChemAlert Storage Symbol White
CAS 7647-01-0
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Molecular Formula HCl·H2O
MDL Number MFCD00011324
Concentration 2.96 to 3.03N
Identification Pass Test
Packaging PolyPac™
Vapor Pressure 5.7mmHg at 0°C
NIST Traceable to NIST.
Melting Point −74°C
Boiling Point 81.5°C
Safety and Handling