missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric Acid, 20% (v/v) Aqueous Solution, Ricca Chemical

Manufacturer:  Ricca Chemical Company RI357432

 View more versions of this product

Catalog No. 3574-32



Description

Description

Specifications

Specifications

Hydrochloric Acid
Colorless to Green-Yellow
7647-01-0,7732-18-5
83.0,19.48
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt
Cl
36.458
CHEBI:17883
Poly natural
20% (v/v)
1L
79.0,18.71
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
313
Liquid
Aqueous Solution

Chemical Identifiers

7647-01-0
36.458
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt
CHEBI:17883
Cl
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
chlorane
SDS
Documents