missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric Acid, 20% (v/v) Aqueous Solution, Ricca Chemical

$72.03 - $131.18

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.458
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym acide chlorhydrique
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name chlorane
SMILES Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
3574-32 View Documents Ricca Chemical Company
RI357432
1L Poly natural Each for $72.03
 
3574-1 View Documents Ricca Chemical Company
RI35741
4L Poly natural Each for $131.18
 
Specifications

Chemical Identifiers

7647-01-0
36.458
acide chlorhydrique
CHEBI:17883
Cl
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
chlorane

Specifications

Colorless to Green-Yellow
7647-01-0,7732-18-5
83.0,19.48
acide chlorhydrique
Cl
36.458
CHEBI:17883
Poly natural
Aqueous Solution
1L
79.0,18.71
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
313
Liquid
20% (v/v)
Hydrochloric Acid