missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydrochloric Acid, 2% (v/v) Aqueous Solution, Ricca Chemical
Manufacturer: Ricca Chemical Company RI356732
Specifications
Hydrochloric Acid | |
Colorless | |
7647-01-0,7732-18-5 | |
100.0,2.35 | |
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt | |
Cl | |
36.458 | |
CHEBI:17883 | |
Poly natural |
2% (v/v) | |
1L | |
96.0,2.26 | |
ClH | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
Liquid | |
Aqueous Solution |
Chemical Identifiers
7647-01-0 | |
36.458 | |
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt | |
CHEBI:17883 | |
Cl |
ClH | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
chlorane |
Promotions
-
Fisherbrand
Spend $250, Get $250 on Ricca Chemical Products
Buy $250 of Ricca Chemical Company products and get $250 more to spend on Ricca items at no additional cost.: 06/30/21