Please login to your online account to display your discounted pricing

Hydrochloric Acid, 2.50 Normal, Ricca Chemical


Quantity 10L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR371500010F

 View more versions of this product

Catalog No. R371500010F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless to slight greenish yellow
Quantity 10L
Chemical Name or Material Hydrochloric Acid
CAS 7732-18-5
CAS Max % 82.0
CAS 7647-01-0
CAS Min % 19.31
CAS Max % 20.1
CAS Min % 79.0
Packaging Cubitainer

Specifications

Chemical Name or Material Hydrochloric Acid
CAS 7732-18-5
CAS Min % 79.0
CAS Max % 82.0
CAS 7647-01-0
CAS Min % 19.31
CAS Max % 20.1
Physical Form Liquid