missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydrochloric Acid, 2.00 N, Ricca Chemical
$66.56 - $633.44
Chemical Identifiers
CAS | 7647-01-0 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid |
PubChem CID | 313 |
ChEBI | CHEBI:23116 |
IUPAC Name | hydrogen chloride |
SMILES | Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
3710-16
|
Ricca Chemical Company
371016 |
500 mL | Natural Poly Bottle |
Each for $66.56
|
|
||||
3710-32
|
Ricca Chemical Company
371032 |
1 L | Natural Poly Bottle |
Each for $99.06
|
|
||||
3710-1
|
Ricca Chemical Company
37101 |
4 L | Natural Poly Bottle |
Each for $305.53
|
|
||||
37102-5
|
Ricca Chemical Company
371025 |
10 L | Cubitainer |
Each for $433.97
|
|
||||
3710-5
|
Ricca Chemical Company
37105 |
20 L | Cubitainer |
Each for $633.44
|
|
||||
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
hydrogen chloride |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid | |
CHEBI:23116 | |
Cl |
Specifications
Colorless | |
16.05,82.0 | |
16.71,85.0 | |
500 mL | |
Liquid | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid | |
Cl | |
36.46 | |
CHEBI:23116 |
7647-01-0,7732-18-5 | |
2.00N | |
Natural Poly Bottle | |
ClH | |
MFCD00011324 MFCD00792839 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 | |
Hydrochloric Acid |