missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric Acid, 1% (v/v) Aqueous Solution, Ricca Chemical

Manufacturer:  Ricca Chemical Company 356032

 View more versions of this product

Catalog No. 3560-32



Description

Description

Specifications

Specifications

1L
97.0,1.14
ClH
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt
[H+].[Cl-]
36.46
CHEBI:17883
7647-01-0,7732-18-5
100.0,1.19
MFCD00011324
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
Poly bottle

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt
CHEBI:17883
[H+].[Cl-]
SDS
Documents