missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydrochloric Acid, 1% (v/v) Aqueous Solution, Ricca Chemical
Manufacturer: Ricca Chemical Company RI356016
Specifications
500mL | |
97.0,1.14 | |
ClH | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
Poly natural |
7647-01-0,7732-18-5 | |
100.0,1.19 | |
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt | |
Cl | |
36.458 | |
CHEBI:17883 |
Chemical Identifiers
7647-01-0 | |
36.458 | |
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt | |
CHEBI:17883 | |
Cl |
ClH | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
chlorane |