missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Hydrochloric Acid, 1% (v/v) Aqueous Solution, Ricca Chemical
Chemical Identifiers
CAS |
7647-01-0 |
Molecular Formula |
ClH |
Molecular Weight (g/mol) |
36.458 |
InChI Key |
VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym |
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt |
PubChem CID |
313 |
ChEBI |
CHEBI:17883 |
IUPAC Name |
chlorane |
SMILES |
Cl |
View More Specs
Products
2
Catalog Number |
Mfr. No. |
Quantity |
Packaging |
Price |
Quantity |
|
|
|
|
3560-16
View Documents
|
Ricca Chemical Company
RI356016
|
500mL |
Poly natural |
Each for $37.09
|
|
Add to cart
|
|
|
3560-32
View Documents
|
Ricca Chemical Company
RI356032
|
1L |
Poly bottle |
Each for $46.15
|
|
Add to cart
|
|
|
Description
Specifications
Chemical Identifiers
|
7647-01-0 |
|
36.458 |
|
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt |
|
CHEBI:17883 |
|
Cl |
|
ClH |
|
VEXZGXHMUGYJMC-UHFFFAOYSA-N |
|
313 |
|
chlorane |
Specifications
|
500mL |
|
97.0,1.14 |
|
ClH |
|
VEXZGXHMUGYJMC-UHFFFAOYSA-N |
|
chlorane |
|
313 |
|
Poly natural |
|
7647-01-0,7732-18-5 |
|
100.0,1.19 |
|
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt |
|
Cl |
|
36.458 |
|
CHEBI:17883 |
SDS