Please login to your online account to display your discounted pricing

Hydrochloric Acid, 1.96% (v/v) Aqueous Solution (1 + 50), Ricca Chemical

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 7732-18-5
CAS Min % 96.0
CAS Max % 100.0
CAS 7647-01-0
CAS Min % 2.22
CAS Max % 2.31
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

3565-9-16

 
ricca chemical company
RI3565.916
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

3565-9-32

 
ricca chemical company
RI3565.932
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R356590020F

 
ricca chemical company
RIR356590020F
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R35659004A

 
ricca chemical company
RIR35659004A
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Hydrochloric Acid
Shelf Life 24 months
Color Clear colorless
Quantity 500mL
CAS 7732-18-5
CAS Min % 96.0
CAS Max % 100.0
CAS 7647-01-0
CAS Min % 2.22
CAS Max % 2.31
Physical Form Liquid
Concentration or Composition (by Analyte or Components) 1.96% (v/v) Aqueous Solution (1 + 50)
Packaging Poly natural
Safety and Handling

24 months