Please login to your online account to display your discounted pricing

Hydrochloric Acid, 1.5% (v/v) Aqueous Solution, Ricca Chemical

$54.70 - $54.70

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 7732-18-5
CAS Min % 96.0
CAS Max % 100.0
CAS 7647-01-0
CAS Min % 1.7
CAS Max % 1.77
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

3565-32

 
ricca chemical company
RI356532
Poly natural 1L Each for $54.70

3565-1

 
ricca chemical company
RI35651
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

3565-16

 
ricca chemical company
RI356516
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R356500020F

 
ricca chemical company
RIR356500020F
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 1L
Concentration or Composition Notes 1.5% (v/v)
Solvent or Matrix Aqueous Solution
CAS Min % 96.0
CAS Max % 100.0
CAS 7732-18-5
Packaging Poly natural
CAS Min % 1.7
CAS Max % 1.77
CAS 7647-01-0
Chemical Name or Material Hydrochloric Acid