Please login to your online account to display your discounted pricing

Hydrochloric Acid (1+3), Fisher Chemical

$73.91 - $73.91

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid (1+3)
CAS 7732-18-5
CAS Min % 76.0
CAS Max % 79.0
CAS 7647-01-0
CAS Min % 22.32
CAS Max % 23.24
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SA9225-1

  SDS
ricca chemical company
SA92251
Poly Bottle 1L Each for $73.91

SA9225-4

  SDS
ricca chemical company
SA92254
Poly Bottle 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SA9225-500

 
ricca chemical company
SA9225500
Poly Bottle 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Hydrochloric Acid (1+3)
Quantity 1L
CAS 7732-18-5
ChemAlert Storage Symbol White
CAS Min % 76.0
CAS Max % 79.0
CAS 7647-01-0
CAS Min % 22.32
CAS Max % 23.24
DOT Information DOT Class 8, : Corrosive
Packaging Poly Bottle
Methods For AOAC Method 960.27
NIST Traceable to NIST.
Safety and Handling