missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chemical Identifiers
CAS | 7647-01-0 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.458 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
IUPAC Name | chlorane |
SMILES | Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
37-001-6CS
|
Ricca Chemical Company
RIR370000012X500 |
6L | Poly natural |
Each for $410.94
|
|
37-003-2CS
|
Ricca Chemical Company
RIR370000012X1A |
12L | Poly natural |
Each for $493.12
|
|
37-001-CS
|
Ricca Chemical Company
RIR37000004X4A |
16L | Poly natural |
Each for $363.94
|
|
Chemical Identifiers
7647-01-0 | |
36.458 | |
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt | |
CHEBI:17883 | |
Cl |
ClH | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
chlorane |
Specifications
6L | |
1.000 ± 0.001 N at 20°C | |
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt | |
Cl | |
36.458 | |
CHEBI:17883 | |
Poly natural | |
Hydrochloric Acid, 1 N |
7647-01-0,7732-18-5 | |
ClH | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
Odorless to slightly pungent | |
Aqueous Solution |