Please login to your online account to display your discounted pricing

Hydrochloric acid, 0.5N Standardized Solution, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Hydrochloric acid
CAS 7647-01-0
CAS 7732-18-5
Physical Form Liquid
Molecular Formula HCl
Formula Weight 36.46
MDL Number MFCD00011324
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA35642K2

 
alfa aesar
35642K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA35642K7

 
alfa aesar
35642K7
4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA35642M4

 
alfa aesar
35642M4
20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 36.46
Color Clear colorless
Quantity 1L
Chemical Name or Material Hydrochloric acid
EINECS Number 231-595-7
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Molecular Formula HCl
MDL Number MFCD00011324
CAS 7647-01-0
Concentration or Composition (by Analyte or Components) Hydrochloric acid: 4.13%; Water: 95.87%
CAS 7732-18-5
Appearance Colorless liquid