Please login to your online account to display your discounted pricing

Hydrochloric Acid, 0.5167 Normal, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
Name Note 0.5167 Normal
CAS 7732-18-5
CAS 7647-01-0
Physical Form Liquid
Assay Percent Range 0.5167 ± 0.0005 N at 20°C
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R36510001A

 
ricca chemical company
RIR36510001A
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R3651000500

 
ricca chemical company
RIR3651000500A
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 1L
Chemical Name or Material Hydrochloric Acid
CAS 7732-18-5
Solvent or Matrix Aqueous Solution
Name Note 0.5167 Normal
Assay Percent Range 0.5167 ± 0.0005 N at 20°C
CAS 7647-01-0
Packaging Poly natural