Please login to your online account to display your discounted pricing

Hydrochloric Acid, 0.5165 Normal, Ricca Chemical

$99.64 - $261.36

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 7647-01-0
CAS Min % 4.69
CAS Max % 4.88
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R365020020F

 
ricca chemical company
RIR365020020F
Cubitainer 20L Each for $261.36

R36502004A

  SDS
ricca chemical company
RIR36502004A
Poly natural 4L Each for $99.64
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 20L
Concentration or Composition Notes 0.5165Normal
Packaging Cubitainer
CAS Min % 4.69
CAS Min % 93.0
CAS Max % 97.0
CAS Max % 4.88
Chemical Name or Material Hydrochloric Acid
CAS 7732-18-5
CAS 7647-01-0