missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydrochloric Acid, 0.300 N, Ricca Chemical
Supplier: Ricca Chemical Company 36305
Specifications
Hydrochloric Acid | |
95.0,2.8 | |
Cubitainer | |
ClH | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
[H+].[Cl-] | |
36.46 | |
CHEBI:17883 |
7647-01-0,7647-01-0 | |
99.0,2.91 | |
20 L | |
MFCD00011324 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 |