Please login to your online account to display your discounted pricing

Hydrochloric Acid, 0.2N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$83.95 - $175.57

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 7647-01-0
Name Note 0.2N Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula HCl
Formula Weight 36.46
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003030

 
avantor performance
561202
Narrow Mouth Polyethylene Bottle 1L Each for $83.95

02003031

 
avantor performance
561203
Cubetainer 4L Each for $175.57
Description & Specifications

Specifications

Formula Weight 36.46
CAS 7647-01-0
Chemical Name or Material Hydrochloric Acid
Molecular Formula HCl
Specific Gravity 1L = 1.01kg
Purity Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Polyethylene Bottle
Name Note 0.2N Volumetric Solution
Quantity 1L