Please login to your online account to display your discounted pricing

Hydrochloric Acid, 0.200 Normal, Ricca Chemical

$40.24 - $193.70

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 7732-18-5
CAS Min % 96.0
CAS Max % 100.0
CAS 7647-01-0
CAS Min % 1.88
CAS Max % 1.96
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

3620-1

  SDS
ricca chemical company
RI36201
Poly natural 4L Each for $88.61

3620-32

  SDS
ricca chemical company
RI362032
Poly natural 1L Each for $40.24

36202-5

 
ricca chemical company
RI36202.5
Cubitainer 10L Each for $193.70

R3620000500

 
ricca chemical company
RIR3620000500A
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Hydrochloric Acid
Color Clear colorless
Quantity 4L
CAS 7732-18-5
CAS Min % 96.0
CAS Max % 100.0
CAS 7647-01-0
CAS Min % 1.88
CAS Max % 1.96
Physical Form Liquid
Packaging Poly natural
Concentration or Composition Notes 0.200Normal