Please login to your online account to display your discounted pricing

Hydrochloric Acid, 0.1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$82.60 - $496.02

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 7647-01-0
Name Note 0.1N Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula HCl
Formula Weight 36.46
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003027

 
avantor performance
562102
Narrow Mouth Poly Bottle 1L Each for $82.60

02003028

 
avantor performance
562103
Cubetainer 4L Each for $496.02
Description & Specifications

Specifications

Formula Weight 36.46
Chemical Name or Material Hydrochloric Acid
CAS 7647-01-0
Molecular Formula HCl
Purity Grade BAKER ANALYZED™ Reagent
Specific Gravity 1L = 0.00kg
Name Note 0.1N Volumetric Solution
Packaging Narrow Mouth Poly Bottle
Quantity 1L