missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric acid, 0.1N Standardized Solution, Alfa Aesar™

$32.01 - $149.46

Chemical Identifiers

CAS 7647-01-0,7732-18-5
Physical Form Liquid
Molecular Formula HCl
Formula Weight 36.46
MDL Number MFCD00011324
Chemical Name or Material Hydrochloric acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA35644K2 SDS Alfa Aesar
35644K2
1L Each for $32.01
AA35644M4 SDS Alfa Aesar
35644M4
20L Each for $149.46
AA35644K7 SDS Alfa Aesar
35644K7
4L Each for $64.03
Specifications Description & Specifications

Chemical Identifiers

7647-01-0,7732-18-5
HCl
MFCD00011324
Liquid
36.46
Hydrochloric acid

Specifications

7647-01-0,7732-18-5
Liquid
36.46
Hydrochloric acid: 0.826%; Water: 99.174%
Clear colorless
Hydrochloric acid
Colorless liquid
HCl
MFCD00011324
23 hPa (17mm Hg) at 20°C
1L

Safety and Handling

EINECSNumber : 231-595-7