Learn More
Hydrochloric acid, 0.1N Standardized Solution, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 035644K2
Description
Hydrochloric acid, 0.1N standardized solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hydrochloric acid | |
1 L | |
ClH | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 | |
36.46 | |
23 hPa (17mm Hg) at 20°C | |
Liquid |
7647-01-0,7732-18-5 | |
Colorless | |
MFCD00011324 MFCD00792839 | |
Cl | |
36.46 | |
CHEBI:17883 | |
Hydrochloric acid: 0.826%; Water: 99.174% | |
Miscible with water, methanol,ethanol and ether |
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
CHEBI:17883 | |
Cl |
ClH | |
MFCD00011324 MFCD00792839 | |
313 | |
hydrogen chloride |
Safety and Handling
P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390
H290-H315-H319
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 231-595-7
RTECSNumber : MW4025000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only