Please login to your online account to display your discounted pricing

Hydrochloric Acid 0.1N DILUT-IT™ Dissolution Media Concentrate, J.T.Baker™

$732.33 - $957.67

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid 0.1N DILUT-IT™ Dissolution Media Concentrate
CAS 7647-01-0
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002963

 
avantor performance
D01104
Narrow Mouth Polyethylene Bottle 1.9L Each for $957.67

02002962

 
avantor performance
D01100
Poly Bottle 230.8mL Each for $732.33
Description & Specifications

Specifications

Chemical Name or Material Hydrochloric Acid 0.1N DILUT-IT™ Dissolution Media Concentrate
Specific Gravity 1L = 1.00kg
CAS 7647-01-0
Packaging Narrow Mouth Polyethylene Bottle
Quantity 1.9L