Please login to your online account to display your discounted pricing

Hydrochloric Acid, 0.1782 Normal, 1mL = 5mg CaO (Lime), Ricca Chemical

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 7732-18-5
CAS Min % 96.0
CAS Max % 100.0
CAS 7647-01-0
CAS Min % 1.68
CAS Max % 1.75
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

3618-1

 
ricca chemical company
RI36181
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

3618-5

 
ricca chemical company
RI36185
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R361800010F

 
ricca chemical company
RIR361800010F
Cubitainer 10L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R36180004F

 
ricca chemical company
RIR36180004F
Cubitainer 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 4L
Concentration or Composition Notes 0.1782Normal
Equivalence 1 mL = 5 mg CaO (Lime)
Chemical Name or Material Hydrochloric Acid
CAS 7732-18-5
CAS Min % 96.0
CAS Max % 100.0
CAS 7647-01-0
CAS Min % 1.68
CAS Max % 1.75
Packaging Poly natural