Please login to your online account to display your discounted pricing

Hydrochloric Acid, 0.1782 Normal, 1mL = 5mg CaO (Lime), Ricca Chemical


Quantity 4L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR36180004F

 View more versions of this product

Catalog No. R36180004F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
Color Clear colorless
Quantity 4L
CAS 7732-18-5
CAS Min % 96.0
CAS Max % 100.0
CAS 7647-01-0
CAS Min % 1.68
CAS Max % 1.75
Physical Form Liquid
Packaging Cubitainer
Concentration or Composition Notes 0.1782Normal
Equivalence 1 mL = 5 mg CaO (Lime)

Specifications

Chemical Name or Material Hydrochloric Acid
CAS 7732-18-5
CAS Min % 96.0
CAS Max % 100.0
CAS 7647-01-0
CAS Min % 1.68
CAS Max % 1.75
Physical Form Liquid