missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric Acid, 0.1782 Normal, 1mL = 5mg CaO (Lime), Ricca Chemical

Manufacturer:  Ricca Chemical Company RIR361800010F

 View more versions of this product

Catalog No. R361800010F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Hydrochloric Acid
10L
96.0,1.68
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
313
Liquid
0.1782Normal
Colorless
7647-01-0,7732-18-5
100.0,1.75
acide chlorhydrique
Cl
36.458
CHEBI:17883
Cubitainer
1 mL = 5 mg CaO (Lime)

Chemical Identifiers

7647-01-0
36.458
acide chlorhydrique
CHEBI:17883
Cl
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
chlorane
Documents