missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric Acid, 0.100 N (N/10), Ricca Chemical

Manufacturer:  Ricca Chemical Company 36005

 View more versions of this product

Catalog No. 3600-5



Description

Description

Specifications

Specifications

Hydrochloric Acid
7647-01-0,7732-18-5
100.0,0.99
MFCD00011324
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
Cubitainer
0.100N
97.0,0.95
ClH
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt
[H+].[Cl-]
36.46
CHEBI:17883
20 L

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt
CHEBI:17883
[H+].[Cl-]
SDS Documents
Product Certifications
Promotions

Promotions