missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric Acid, 0.100 Normal, Ricca Chemical

$347.06 - $483.97

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES [H+].[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
36-003-2CS
View Documents Promotion Details
Ricca Chemical Company
R360000012X1A
12L Poly natural
Each for $483.97
 
36-001-CS
View Documents Promotion Details
Ricca Chemical Company
R36000004X4A
16L Poly natural
Each for $347.06
 
Description

Description

Specifications

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt
CHEBI:17883
[H+].[Cl-]

Specifications

Colorless
7647-01-0,7732-18-5
ClH
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt
[H+].[Cl-]
36.46
CHEBI:17883
Odorless to slightly pungent
Aqueous Solution
12L
0.1000 ± 0.0001 N at 20°C
MFCD00011324
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
Liquid
Poly natural
Hydrochloric Acid, 0.100 Normal
SDS
Product Certifications