missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric Acid, 0.100 Normal, Ricca Chemical

Manufacturer:  Ricca Chemical Company RIR360000012X1A

 View more versions of this product

Catalog No. 36-003-2CS



Description

Description

Specifications

Specifications

Hydrochloric Acid
12L
7647-01-0,7732-18-5
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
313
Liquid
Poly natural
Colorless
0.100 Normal
0.1000 ± 0.0001 N at 20°C
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt
Cl
36.458
CHEBI:17883
Odorless to slightly pungent
Aqueous Solution

Chemical Identifiers

7647-01-0
36.458
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt
CHEBI:17883
Cl
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
chlorane
SDS
Documents
Promotions

Promotions