missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric acid, 0.05N Standardized Solution, Alfa Aesar™

$32.64 - $32.64

Chemical Identifiers

CAS 7647-01-0,7732-18-5
Physical Form Liquid
Molecular Formula HCl
Formula Weight 36.46
MDL Number MFCD00011324
Chemical Name or Material Hydrochloric acid, 0.05N
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA35646K2 SDS Alfa Aesar
35646K2
1L Each for $32.64
AA35646K7 SDS Alfa Aesar
35646K7
4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA35646M4 SDS Alfa Aesar
35646M4
20L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7647-01-0,7732-18-5
HCl
MFCD00011324
Liquid
36.46
Hydrochloric acid, 0.05N

Specifications

Clear colorless
36.46
7732-18-5,7647-01-0
MFCD00011324
Colorless liquid
Hydrochloric acid, 0.05N
Liquid
1L
Hydrochloric acid: 0.413%; Water: 99.587%
HCl
23 hPa (17mm Hg) at 20°C

Safety and Handling

EINECSNumber : 231-595-7