missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric Acid, 0.0200 Normal, Ricca Chemical

$54.21 - $106.33

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.458
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name chlorane
SMILES Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
3595-32
View Documents
Ricca Chemical Company
RI359532
1L Poly natural
Each for $54.21
 
36-103-1120
View Documents
Ricca Chemical Company
35951
4L Poly natural
Each for $106.33
 
R35950004F
View Documents
Ricca Chemical Company
RIR35950004F
4L Cubitainer
Each for $104.85
 
Description

Description

Specifications

Chemical Identifiers

7647-01-0
36.458
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt
CHEBI:17883
Cl
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
chlorane

Specifications

Colorless
7647-01-0,7732-18-5
100.0,0.2
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt
Cl
36.458
CHEBI:17883
Poly natural
Hydrochloric Acid
1L
98.0,0.19
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
313
Liquid
0.0200Normal
SDS