Please login to your online account to display your discounted pricing

Hydrochloric Acid, 0.01N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$82.44 - $463.87

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 7647-01-0
Name Note 0.01N Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula HCl
Formula Weight 36.46
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003023

 
avantor performance
561102
Narrow Mouth Polyethylene Bottle 1L Each for $82.44

02003024

 
avantor performance
561103
Cubetainer 4L Each for $463.87
Description & Specifications

Specifications

Formula Weight 36.46
Chemical Name or Material Hydrochloric Acid
Molecular Formula HCl
CAS 7647-01-0
Specific Gravity 1L = 1.00kg
Name Note 0.01N Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Polyethylene Bottle
Quantity 1L