Please login to your online account to display your discounted pricing

Hydrochloric acid, 0.01N Standardized Solution, Alfa Aesar™

$63.28 - $63.28

Chemical Identifiers

Chemical Name or Material Hydrochloric acid
Name Note 0.01N
CAS 7647-01-0
CAS 7732-18-5
Physical Form Liquid
Molecular Formula HCl
Formula Weight 36.46
MDL Number MFCD00011324
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA35648K2

  SDS
alfa aesar
35648K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA35648K7

  SDS
alfa aesar
35648K7
4L Each for $63.28

AA35648M4

  SDS
alfa aesar
35648M4
20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Formula Weight 36.46
Physical Form Liquid
Quantity 1L
EINECS Number 231-595-7
Concentration or Composition (by Analyte or Components) Hydrochloric acid: 0.083%; Water: 99.917%
Name Note 0.01N
Vapor Pressure 23 hPa (17mm Hg) at 20°C
CAS 7647-01-0
CAS 7732-18-5
Molecular Formula HCl
Appearance Colorless liquid
MDL Number MFCD00011324
Chemical Name or Material Hydrochloric acid