Please login to your online account to display your discounted pricing

Hydrochloric Acid 0.01N DILUT-IT™ Dissolution Media Concentrate, J.T.Baker™

$725.41 - $1,008.30

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid 0.01N DILUT-IT™ Dissolution Media Concentrate
CAS 7647-01-0
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002960

 
avantor performance
D01000
Poly Bottle 230.8mL Each for $725.41

02002961

 
avantor performance
D01004
Narrow Mouth Polyethylene Bottle 1.9L Each for $1,008.30
Description & Specifications

Specifications

CAS 7647-01-0
Chemical Name or Material Hydrochloric Acid 0.01N DILUT-IT™ Dissolution Media Concentrate
Specific Gravity 1L = 1.00kg
Packaging Poly Bottle
Quantity 230.8mL