missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric Acid, 0.0100 Normal, Ricca Chemical

$34.33 - $34.33

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.458
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym acide chlorhydrique
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name chlorane
SMILES Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
R3590000500 View Documents Ricca Chemical Company
RIR3590000500A
500mL Poly natural Each for $34.33
 
R35900004F View Documents Ricca Chemical Company
RIR35900004F
4L Cubitainer N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

7647-01-0
36.458
acide chlorhydrique
CHEBI:17883
Cl
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
chlorane

Specifications

Colorless
7647-01-0,7732-18-5
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
313
Liquid
Odorless to slightly pungent
Solubility in Water: Infinite
Hydrochloric Acid, 0.0100 Normal
500mL
0.01000 ± 0.00005 N at 20°C
acide chlorhydrique
Cl
36.458
CHEBI:17883
ACS
Poly natural
Aqueous Solution