missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ Hydrobromic Acid Solution, purum p.a., ≥62% (T), Honeywell™

Purum p.a.,≥62% (T)

$153.04 - $153.04

Chemical Identifiers

CAS 10035-10-6
Molecular Formula BrH
Molecular Weight (g/mol) 80.912
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
IUPAC Name bromane
SMILES Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60045067 View Documents Honeywell™
18720250ML
250mL Glass Bottle Each for $153.04
 
Description

Description

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications

Chemical Identifiers

10035-10-6
80.912
CPELXLSAUQHCOX-UHFFFAOYSA-N
260
bromane
BrH
MFCD00011323
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
CHEBI:47266
Br

Specifications

Brown-Yellow
10035-10-6
MFCD00011323
3587158
CPELXLSAUQHCOX-UHFFFAOYSA-N
bromane
80.912
CHEBI:47266
≥62% (T)
Glass Bottle
-11°C
Hydrobromic Acid Solution
250mL
BrH
UN1788
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
Br
1.73g/cu.cm
260
80.91g/mol
purum p.a.
Clear liquid
126°C
Safety and Handling

Safety and Handling

P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338

H314-H335

DOTInformation : 8/PGII