missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrobromic acid, Alfa Aesar™

Manufacturer:  Alfa Aesar 14036AT

 View more versions of this product

Catalog No. AA14036AT



Specifications

Specifications

Hydrobromic acid
10035-10-6
MFCD00011323
CPELXLSAUQHCOX-UHFFFAOYSA-N
bromane
80.912
CHEBI:47266
Liquid
940mL
BrH
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
Br
1.49g/mL
260
80.92

Chemical Identifiers

10035-10-6
80.912
CPELXLSAUQHCOX-UHFFFAOYSA-N
260
bromane
BrH
MFCD00011323
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
CHEBI:47266
Br
Safety and Handling

Safety and Handling

EINECSNumber : 233-113-0

Documents