Learn More
Hydrobromic acid, ACS, 47.0-49.0%, Thermo Scientific Chemicals
Air Sensitive
Supplier: Thermo Scientific Chemicals 036694M1
Description
Hydrobromic acid is an indispensable raw material for organic intermediates, inorganic pharmaceuticals, photosensitive materials, dyes, and medicines. It is also used in the production of bromine compounds. It is used as an important agent in the industrial processes of isomerization, polymerization, hydration, and dehydration, and esterification reactions. It plays an essential role for the extraction of mercury from the cinnebar ore.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Colorless to pale yellow liquid | |
Colorless to Yellow | |
1.49 | |
22500 hPa (16876mm Hg) at 20°C | |
4 x 1 L | |
Liquid | |
3587158 | |
Miscible in water. | |
Br | |
80.91 | |
CHEBI:47266 | |
ACS |
Hydrobromic acid | |
10035-10-6,7732-18-5 | |
Air Sensitive | |
47.0-49.0% | |
BrH | |
MFCD00011323 | |
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide | |
CPELXLSAUQHCOX-UHFFFAOYSA-N | |
hydrogen bromide | |
260 | |
80.92 | |
Hydrogen bromide: 48.0%; Water: 52.0% |
Chemical Identifiers
10035-10-6 | |
80.91 | |
CPELXLSAUQHCOX-UHFFFAOYSA-N | |
260 | |
Br |
BrH | |
MFCD00011323 | |
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide | |
CHEBI:47266 |
Safety and Handling
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : Hazard Class: 8, Packing Group: II
EINECSNumber : 233-113-0
RTECSNumber : MW3850000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only