missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydrobenzamide, 98%

Manufacturer:  Alfa Aesar™ H5349214

 View more versions of this product

Catalog No. AAH5349214



Specifications

Specifications

Hydrobenzamide
25g
C21H18N2
UN2811
hydrobenzamid
C1=CC=C(C=C1)C=NC(C2=CC=CC=C2)N=CC3=CC=CC=C3
298.389
298.39
102°C to 105°C
92-29-5
MFCD00022039
615231
VUYRFIIFTJICNA-LKNRODPVSA-N
(E)-N-[[(E)-benzylideneamino]-phenylmethyl]-1-phenylmethanimine
9566477
98%

Chemical Identifiers

92-29-5
298.389
VUYRFIIFTJICNA-LKNRODPVSA-N
9566477
C1=CC=C(C=C1)C=NC(C2=CC=CC=C2)N=CC3=CC=CC=C3
C21H18N2
MFCD00022039
hydrobenzamid
(E)-N-[[(E)-benzylideneamino]-phenylmethyl]-1-phenylmethanimine
Safety and Handling

Safety and Handling

GHS H Statement
H300
Fatal if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA

Documents