missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydrobenzamide, 98%

Manufacturer:  Alfa Aesar™ H5349206

 View more versions of this product

Catalog No. AAH5349206



Specifications

Specifications

Hydrobenzamide
5g
C21H18N2
UN2811
hydrobenzamid
C1=CC=C(C=C1)C=NC(C2=CC=CC=C2)N=CC3=CC=CC=C3
298.389
298.39
102°C to 105°C
92-29-5
MFCD00022039
615231
VUYRFIIFTJICNA-LKNRODPVSA-N
(E)-N-[[(E)-benzylideneamino]-phenylmethyl]-1-phenylmethanimine
9566477
98%

Chemical Identifiers

92-29-5
298.389
VUYRFIIFTJICNA-LKNRODPVSA-N
9566477
C1=CC=C(C=C1)C=NC(C2=CC=CC=C2)N=CC3=CC=CC=C3
C21H18N2
MFCD00022039
hydrobenzamid
(E)-N-[[(E)-benzylideneamino]-phenylmethyl]-1-phenylmethanimine
Safety and Handling

Safety and Handling

GHS H Statement
H300
Fatal if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 202-144-1

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents