Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydrindantin dihydrate, 98%

$77.22 - $262.61

Chemical Identifiers

Chemical Name or Material Hydrindantin Dihydrate
CAS 5950-69-6
Molecular Formula C18H10O6·2H2O
Formula Weight 358.30g/mol (322.27g/mol Anhydrous)
MDL Number MFCD00149242
Beilstein 1895666
Merck Index 14,4775
Synonym 2,2',3,3,3',3'-Hexahydroxy-2,2'-biindan-1,1'-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1246214

 
alfa aesar™
A12462-14
25g Each for $77.22

AAA1246222

 
alfa aesar™
A12462-22
100g Each for $262.61
Description & Specifications

Specifications

Melting Point ≈250°C (dec.)
Quantity 25g
Chemical Name or Material Hydrindantin dihydrate
CAS 5950-69-6
Assay 98%
Molecular Formula C18H10O6·2H2O
Formula Weight 358.30g/mol (322.27g/mol Anhydrous)
MDL Number MFCD00149242
Beilstein 1895666
Merck Index 14,4775
Synonym 2,2',3,3,3',3'-Hexahydroxy-2,2'-biindan-1,1'-dione
RTECS Number DU2989000
TSCA TSCA

Applications
Hydrindantin dihydrate is a reagent for photometric determination of amino acids and also used in amino acid determinations as a ninhydrin synergist. It is a chemical used in Stein-Moore-Chromatography.

Solubility
Insoluble in water. Soluble with decomposition in aqueous sodium carbonate (deep red color) and in sodium hydroxide solutions (deep blue color).

Notes
Store under cool dry place. Ensure good ventilation. Incompatible with oxidizing agents.