missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydridotetrakis(triphenylphosphine)rhodium(I), Rh 8.9% min

Manufacturer:  Alfa Aesar™ 4403006

 View more versions of this product

Catalog No. AA4403006



Specifications

Specifications

Hydridotetrakis (triphenylphosphine)rhodium (I)
145°C to 147°C (decomposition)
18284-36-1
MFCD00015867
Rh 8.9% min
IGTVAJJAPJKARK-UHFFFAOYSA-N
rhodium;triphenylphosphane
11981874
Powder
Air Sensitive
5g
C72H60P4Rh
cas number:73468-85-6
Soluble in chloroform, toluene
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Rh]
1152.073
1153.09

Chemical Identifiers

18284-36-1
1152.073
IGTVAJJAPJKARK-UHFFFAOYSA-N
11981874
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Rh]
C72H60P4Rh
MFCD00015867
cas number:73468-85-6
rhodium;triphenylphosphane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents