missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydridotetrakis(triphenylphosphine)rhodium(I), Rh 8.9% min

$422.50 - $1,589.07

Chemical Identifiers

CAS 18284-36-1
Molecular Formula C72H60P4Rh
Molecular Weight (g/mol) 1152.073
MDL Number MFCD00015867
InChI Key IGTVAJJAPJKARK-UHFFFAOYSA-N
Synonym cas number:73468-85-6, hrh pph3 4, hydridotetrakis triphenylphosphine rhodium, hydridotetrakis triphenylphosphine rhodium i, molecular formula:c16h26o2rh2, molecular weight:456.19, rhodium, tetrakis triphenylphosphine, tetrakis triphenylphosphine rhodium, tetrakis triphenylphosphine rhodium hydride, tetrakis triphenylphosphine rhodium i hydride
PubChem CID 11981874
IUPAC Name rhodium;triphenylphosphane
SMILES C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Rh]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4403003 View Documents Alfa Aesar™
4403003
1g Each for $422.50
 
AA4403006 View Documents Alfa Aesar™
4403006
5g Each for $1,589.07
 
Specifications

Chemical Identifiers

18284-36-1
1152.073
IGTVAJJAPJKARK-UHFFFAOYSA-N
11981874
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Rh]
C72H60P4Rh
MFCD00015867
cas number:73468-85-6, hrh pph3 4, hydridotetrakis triphenylphosphine rhodium, hydridotetrakis triphenylphosphine rhodium i, molecular formula:c16h26o2rh2, molecular weight:456.19, rhodium, tetrakis triphenylphosphine, tetrakis triphenylphosphine rhodium, tetrakis triphenylphosphine rhodium hydride, tetrakis triphenylphosphine rhodium i hydride
rhodium;triphenylphosphane

Specifications

Air Sensitive
1g
C72H60P4Rh
cas number:73468-85-6, hrh pph3 4, hydridotetrakis triphenylphosphine rhodium, hydridotetrakis triphenylphosphine rhodium i, molecular formula:c16h26o2rh2, molecular weight:456.19, rhodium, tetrakis triphenylphosphine, tetrakis triphenylphosphine rhodium, tetrakis triphenylphosphine rhodium hydride, tetrakis triphenylphosphine rhodium i hydride
Soluble in chloroform, toluene
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Rh]
1152.073
1153.09
Hydridotetrakis (triphenylphosphine)rhodium (I)
145°C to 147°C (decomposition)
18284-36-1
MFCD00015867
Rh 8.9% min
IGTVAJJAPJKARK-UHFFFAOYSA-N
rhodium;triphenylphosphane
11981874
Powder
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures