Please login to your online account to display your discounted pricing

Hydridotetrakis(triphenylphosphine)rhodium(I), ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC377932500

 View more versions of this product

Catalog No. AC377932500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 1151.25
Quantity 250mg
Color Ochre
Chemical Name or Material Hydridotetrakis(triphenylphosphine)rhodium(I)
Infrared Spectrum Authentic
Molecular Formula C72H61P4Rh
Packaging Glass bottle
CAS 18284-36-1
Rhodium (Rh) 8.5% min.
MDL Number MFCD00015867

Specifications

Chemical Name or Material Hydridotetrakis(triphenylphosphine)rhodium(I)
CAS 18284-36-1
Molecular Formula C72H61P4Rh
Formula Weight 1151.25
MDL Number MFCD00015867