missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrazine sulfate, Reagent ACS, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC423775000

 View more versions of this product

Catalog No. AC423775000


 

Specifications Description & Specifications

Specifications

≥99.0%
Crystalline Powder
500g
15, 4810
0.005% max.
10034-93-2
MFCD00044873
0.05% max.
50ppm max.
130.12
ACS Reagent Grade
White
10ppm max.
H4N2·H2SO4
Hydrazine sulfate
Plastic bottle
20ppm max.
H2NNH2·H2SO4

Chemical Identifiers

10034-93-2
H2NNH2·H2SO4
MFCD00044873
≥99.0%
H4N2·H2SO4
130.12
Hydrazine sulfate
ACS Reagent Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause cancer.
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger