Please login to your online account to display your discounted pricing

Hydrazine sulfate, Reagent ACS, ACROS Organics™


Quantity 50g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  423770500

 View more versions of this product

Catalog No. AC423770500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Formula Weight 130.12
Physical Form Crystalline Powder
Purity Grade ACS Reagent Grade
Quantity 50g
Color White
Merck Index 15, 4810
Heavy Metals (as Pb) 20ppm max.
Iron (Fe) 10ppm max.
Molecular Formula H4N2·H2SO4
Chlorine (Cl) 50ppm max.
Health Hazard 1 Danger
MDL Number MFCD00044873
Residue after Ignition 0.05% max.
Insoluble Matter 0.005% max.
Packaging Plastic bottle
Linear Formula H2NNH2·H2SO4
Chemical Name or Material Hydrazine sulfate
CAS 10034-93-2
Health Hazard 2 GHS H Statement:
May cause cancer.
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Hydrazine sulfate
CAS 10034-93-2
Purity Grade ACS Reagent Grade
Assay ≥99.0%
Molecular Formula H4N2·H2SO4
Linear Formula H2NNH2·H2SO4
Formula Weight 130.12
MDL Number MFCD00044873
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause cancer.
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents