Please login to your online account to display your discounted pricing

Hydrazine Sulfate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$240.59 - $240.59

Chemical Identifiers

Chemical Name or Material Hydrazine Sulfate
CAS 10034-93-2
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula (NH2)2·H2SO4
Formula Weight 130.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003180

 
avantor performance
217701
Poly Bottle 500g Each for $240.59
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 130.12
Physical Form Crystals
Chemical Name or Material Hydrazine Sulfate
CAS 10034-93-2
Molecular Formula (NH2)2·H2SO4
Packaging Poly Bottle
Quantity 500g
Specific Gravity 1L = 1.37kg